siRNA Details
| Gene Symbol: Cfap57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC094257.1 | - | 1 | 242 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AA123023.1 | - | 1 | 233 | Chr. 4 - 118620361 - 118621185 [-] on Build GRCm38 | |

Cfap57
Genomic Map