siRNA Details
| Gene Symbol: Gm2449 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK076661.1 | - | 2 | 57 | ||
| AK132067.1 | - | 2 | 75 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm2449| Gene Symbol: Gm2449 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK076661.1 | - | 2 | 57 | ||
| AK132067.1 | - | 2 | 75 | |||