siRNA Details
| Gene Symbol: Gm17644 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045297.1 | 3 | 2591 | |||
| GenBank mRNA | AK029971.1 | - | 3 | 2592 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm17644| Gene Symbol: Gm17644 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045297.1 | 3 | 2591 | |||
| GenBank mRNA | AK029971.1 | - | 3 | 2592 | ||