siRNA Details
| Gene Symbol: 1110003F10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK003351.1 | - | 1 | 496 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY703608.1 | - | 1 | 494 | Chr. 5 - 65405752 - 65406519 [+] on Build GRCm38 | |

1110003F10Rik
Genomic Map