siRNA Details
| Gene Symbol: 1700047N06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006719.1 | - | 1 | 350 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706708.1 | - | 1 | 350 | Chr. 1 - 10057057 - 10057944 [-] on Build GRCm38 | |

1700047N06Rik
Genomic Map