siRNA Details
| Gene Symbol: 4933417O13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045842.1 | 3 | 317 | |||
| GenBank mRNA | AK016851.1 | - | 3 | 317 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY716526.1 | - | 3 | 317 | Chr. 7 - 143323680 - 143330650 [-] on Build GRCm38 | |

4933417O13Rik
Genomic Map