siRNA Details
| Gene Symbol: 4933417O13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045842.1 | 3 | 1244 | |||
| GenBank mRNA | AK016851.1 | - | 3 | 1244 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933417O13Rik| Gene Symbol: 4933417O13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045842.1 | 3 | 1244 | |||
| GenBank mRNA | AK016851.1 | - | 3 | 1244 | ||