siRNA Details
| Gene Symbol: 1700119I11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018965.1 | - | 3 | 320 | ||
| AV209480.1 | - | 2 | 297 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY707296.1 | - | 3 | 320 | Chr. 13 - 91861432 - 91862056 [+] on Build GRCm38 | |

1700119I11Rik
Genomic Map