siRNA Details
| Gene Symbol: 4930520E11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015663.1 | - | 1 | 719 | ||
| AK015850.1 | - | 1 | 647 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930520E11Rik| Gene Symbol: 4930520E11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015663.1 | - | 1 | 719 | ||
| AK015850.1 | - | 1 | 647 | |||