siRNA Details
| Gene Symbol: Atg12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK080409.1 | - | 3307 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB244840.2 | - | 1 | 423 | Chr. 18 - 46730144 - 46730822 [-] on Build GRCm38 | |

Atg12
Genomic Map