siRNA Details
| Gene Symbol: 1700031L13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006457.1 | - | 2 | 615 | ||
| AK006583.1 | - | 2 | 614 | |||
| AV044120.2 | - | 1 | 110 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CA464362.1 | - | 2 | 619 | Chr. 5 - 82122503 - 82124713 [-] on Build GRCm38 | |

1700031L13Rik
Genomic Map