siRNA Details
| Gene Symbol: 1700028D13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045377.1 | 2 | 253 | |||
| GenBank mRNA | AK006451.1 | - | 2 | 212 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706495.1 | - | 2 | 212 | Chr. 5 - 112206806 - 112210001 [+] on Build GRCm38 | |

1700028D13Rik
Genomic Map