siRNA Details
| Gene Symbol: 1700028D13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045377.1 | 1 | 91 | |||
| NR_045378.1 | 1 | 91 | ||||
| GenBank mRNA | AK006451.1 | - | 1 | 50 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700028D13Rik| Gene Symbol: 1700028D13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045377.1 | 1 | 91 | |||
| NR_045378.1 | 1 | 91 | ||||
| GenBank mRNA | AK006451.1 | - | 1 | 50 | ||