siRNA Details
| Gene Symbol: Gm29811 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102297.1 | 1 | 395 | |||
| NR_102298.1 | 1 | 395 | ||||
| GenBank mRNA | AK135894.1 | - | 1 | 396 | ||
| CA556121.1 | - | 1 | 363 | |||
| CF173244.1 | - | 1 | 385 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm29811| Gene Symbol: Gm29811 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102297.1 | 1 | 395 | |||
| NR_102298.1 | 1 | 395 | ||||
| GenBank mRNA | AK135894.1 | - | 1 | 396 | ||
| CA556121.1 | - | 1 | 363 | |||
| CF173244.1 | - | 1 | 385 | |||