siRNA Details
| Gene Symbol: 9130016M20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018631.1 | - | 1 | 224 | ||
| AK050221.1 | - | 1 | 1369 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY720812.1 | - | 1 | 224 | Chr. 19 - 31951227 - 31952194 [+] on Build GRCm38 | |

9130016M20Rik
Genomic Map