siRNA Details
| Gene Symbol: 2900040C04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013627.1 | - | 1 | 16 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY712593.1 | - | 13 | Chr. 14 - 76251462 - 76252325 [+] on Build GRCm38 | ||

2900040C04Rik
Genomic Map