siRNA Details
| Gene Symbol: Gm7538 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046033.1 | 3 | 1053 | |||
| GenBank mRNA | AK135898.1 | - | 3 | 1054 | ||
| AK162140.1 | - | 3 | 1054 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm7538| Gene Symbol: Gm7538 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046033.1 | 3 | 1053 | |||
| GenBank mRNA | AK135898.1 | - | 3 | 1054 | ||
| AK162140.1 | - | 3 | 1054 | |||