siRNA Details
| Gene Symbol: 4930447E19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019616.1 | - | 1 | 178 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY714955.1 | - | 1 | 178 | Chr. 5 - 101032634 - 101033252 [-] on Build GRCm38 | |

4930447E19Rik
Genomic Map