siRNA Details
| Gene Symbol: 5430409L15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019945.1 | - | 1 | 57 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY717097.1 | - | 1 | 57 | Chr. 11 - 79960112 - 79960519 [-] on Build GRCm38 | |

5430409L15Rik
Genomic Map