siRNA Details
| Gene Symbol: 5430434F05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019948.1 | - | 1 | 214 | ||
| AK162538.1 | - | 1 | 2812 | |||
| Gene Symbol: Grk6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AW048460.1 | - | 1 | 75 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY717202.1 | - | 1 | 213 | Chr. 6 - 83417802 - 83418388 [-] on Build GRCm38 | |

5430434F05Rik
Genomic Map