siRNA Details
| Gene Symbol: Gm10814 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045783.1 | 5 | 1398 | |||
| GenBank mRNA | AK160921.1 | - | 5 | 1399 | ||
| BB114209.2 | - | 2 | 126 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm10814| Gene Symbol: Gm10814 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045783.1 | 5 | 1398 | |||
| GenBank mRNA | AK160921.1 | - | 5 | 1399 | ||
| BB114209.2 | - | 2 | 126 | |||