siRNA Details
Gene Symbol: Gm10814 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045783.1 | 5 | 1896 | |||
GenBank mRNA | AK160921.1 | - | 5 | 1897 | ||
BB114209.2 | - | 2 | 624 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm10814 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045783.1 | 5 | 1896 | |||
GenBank mRNA | AK160921.1 | - | 5 | 1897 | ||
BB114209.2 | - | 2 | 624 |