siRNA Details
| Gene Symbol: 1810014P07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007501.1 | - | 1 | 76 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1810014P07Rik| Gene Symbol: 1810014P07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007501.1 | - | 1 | 76 | ||