siRNA Details
| Gene Symbol: Gm5108 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK076825.1 | - | 4 | 431 | ||
| BC120772.1 | - | 4 | 359 | |||
| BC120798.1 | - | 4 | 359 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5108| Gene Symbol: Gm5108 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK076825.1 | - | 4 | 431 | ||
| BC120772.1 | - | 4 | 359 | |||
| BC120798.1 | - | 4 | 359 | |||