siRNA Details
| Gene Symbol: D830026I12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102304.1 | 2 | 368 | |||
| NR_102305.1 | 2 | 326 | ||||
| GenBank mRNA | AK052888.1 | - | 355 | |||
| AK081964.1 | - | 2 | 369 | |||
| CJ098230.1 | - | 2 | 327 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D830026I12Rik| Gene Symbol: D830026I12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102304.1 | 2 | 368 | |||
| NR_102305.1 | 2 | 326 | ||||
| GenBank mRNA | AK052888.1 | - | 355 | |||
| AK081964.1 | - | 2 | 369 | |||
| CJ098230.1 | - | 2 | 327 | |||