siRNA Details
| Gene Symbol: Gabpb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007509.1 | - | 3 | 214 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gabpb2| Gene Symbol: Gabpb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007509.1 | - | 3 | 214 | ||