siRNA Details
| Gene Symbol: 1700066J03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006903.1 | - | 2 | 495 | ||
| AK018941.1 | - | 1 | 411 | |||
| BY707210.1 | - | 1 | 411 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706900.1 | - | 2 | 491 | Chr. 13 - 58146952 - 58148974 [+] on Build GRCm38 | |

1700066J03Rik
Genomic Map