siRNA Details
| Gene Symbol: Fam219aos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045726.1 | 3 | 670 | |||
| GenBank mRNA | AK006904.1 | - | 3 | 671 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB615121.1 | - | 5 | 600 | Chr. 4 - 41507793 - 41518931 [+] on Build GRCm38 | |

Fam219aos
Genomic Map