siRNA Details
| Gene Symbol: Ccdc138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018096.1 | - | 1 | 37 | ||
| AK043811.1 | - | 4 | 1396 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc138| Gene Symbol: Ccdc138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018096.1 | - | 1 | 37 | ||
| AK043811.1 | - | 4 | 1396 | |||