siRNA Details
| Gene Symbol: Gm6980 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK086943.1 | - | 253 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB605849.2 | - | 3 | 253 | Chr. 9 - 51913800 - 51931661 [-] on Build GRCm38 | |

Gm6980
Genomic Map