siRNA Details
| Gene Symbol: 1810053B23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040487.1 | 1 | 79 | |||
| GenBank mRNA | AK160437.1 | - | 1 | 80 | ||
| BY707451.1 | - | 1 | 80 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1810053B23Rik| Gene Symbol: 1810053B23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040487.1 | 1 | 79 | |||
| GenBank mRNA | AK160437.1 | - | 1 | 80 | ||
| BY707451.1 | - | 1 | 80 | |||