siRNA Details
| Gene Symbol: 1810053B23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040487.1 | 3 | 1262 | |||
| GenBank mRNA | AK160437.1 | - | 3 | 1264 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1810053B23Rik| Gene Symbol: 1810053B23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040487.1 | 3 | 1262 | |||
| GenBank mRNA | AK160437.1 | - | 3 | 1264 | ||