siRNA Details
| Gene Symbol: 2900092N11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013842.1 | - | 1 | 229 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY712809.1 | - | 1 | 229 | Chr. 3 - 80843155 - 80843547 [-] on Build GRCm38 | |

2900092N11Rik
Genomic Map