siRNA Details
| Gene Symbol: Gm11100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK085942.1 | - | 4 | 1510 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm11100| Gene Symbol: Gm11100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK085942.1 | - | 4 | 1510 | ||