siRNA Details
| Gene Symbol: Nkx1-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF222442.1 | - | 1653 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nkx1-2| Gene Symbol: Nkx1-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF222442.1 | - | 1653 | |||