siRNA Details
| Gene Symbol: Gm2090 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK076896.1 | - | 71 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB615660.1 | - | 1 | 71 | Chr. 12 - 87988036 - 88068304 [-] on Build GRCm38 | |

Gm2090
Genomic Map