siRNA Details
| Gene Symbol: Otud3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK076876.1 | - | 1 | 508 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV267369.2 | - | 1 | 102 | Chr. 4 - 138891072 - 138891721 [-] on Build GRCm38 | |

Otud3
Genomic Map