siRNA Details
| Gene Symbol: 6720483E21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040492.1 | 4 | 884 | |||
| GenBank mRNA | AK020171.1 | - | 4 | 889 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

6720483E21Rik| Gene Symbol: 6720483E21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040492.1 | 4 | 884 | |||
| GenBank mRNA | AK020171.1 | - | 4 | 889 | ||