siRNA Details
| Gene Symbol: 6720484G13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020175.1 | - | 1 | 25 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY719464.1 | - | 1 | 25 | Chr. 9 - 101010436 - 101011051 [-] on Build GRCm38 | |

6720484G13Rik
Genomic Map