siRNA Details
| Gene Symbol: Slc4a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK050563.1 | - | 1 | 1665 | ||
| AK083945.1 | - | 1 | 1686 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc4a8| Gene Symbol: Slc4a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK050563.1 | - | 1 | 1665 | ||
| AK083945.1 | - | 1 | 1686 | |||