siRNA Details
Gene Symbol: E330032C10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK143670.1 | - | 3 | 1896 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB736367.1 | - | 3 | 257 | Chr. 17 - 73252628 - 73254146 [+] on Build GRCm38 |
