siRNA Details
Gene Symbol: E330032C10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK143670.1 | - | 1 | 188 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB842860.1 | - | 1 | 189 | Chr. 17 - 73250986 - 73251371 [+] on Build GRCm38 |
