siRNA Details
| Gene Symbol: Gm15408 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040429.1 | 3 | 590 | |||
| GenBank mRNA | AK087624.1 | - | 3 | 322 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB666023.1 | - | 3 | 319 | Chr. 5 - 149007233 - 149012341 [+] on Build GRCm38 | |

Gm15408
Genomic Map