siRNA Details
| Gene Symbol: Gm15408 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040429.1 | 4 | 1210 | |||
| GenBank mRNA | AK087624.1 | - | 4 | 943 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm15408| Gene Symbol: Gm15408 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040429.1 | 4 | 1210 | |||
| GenBank mRNA | AK087624.1 | - | 4 | 943 | ||