siRNA Details
Gene Symbol: Zfp579 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK082494.1 | - | 1333 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB388335.2 | - | 1 | 471 | Chr. 7 - 4983483 - 4985677 [-] on Build GRCm38 |
