siRNA Details
| Gene Symbol: Dmrta2os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019375.1 | - | 2 | 376 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY712842.1 | - | 2 | 376 | Chr. 4 - 109976744 - 109977491 [-] on Build GRCm38 | |

Dmrta2os
Genomic Map