siRNA Details
| Gene Symbol: 4933416O17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006848.1 | - | 2 | 571 | ||
| AK015718.1 | - | 2 | 633 | |||
| AK016836.1 | - | 568 | ||||
| AV204930.1 | - | 1 | 178 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933416O17Rik| Gene Symbol: 4933416O17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006848.1 | - | 2 | 571 | ||
| AK015718.1 | - | 2 | 633 | |||
| AK016836.1 | - | 568 | ||||
| AV204930.1 | - | 1 | 178 | |||