siRNA Details
| Gene Symbol: 4930448L02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019618.1 | - | 1 | 61 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY714969.1 | - | 1 | 61 | Chr. 11 - 30771290 - 30771674 [-] on Build GRCm38 | |

4930448L02Rik
Genomic Map