siRNA Details
| Gene Symbol: Gm4146 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK043255.1 | - | 2 | 260 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB640661.1 | - | 2 | 260 | Chr. 18 - 47761266 - 47789533 [-] on Build GRCm38 | |

Gm4146
Genomic Map