siRNA Details
| Gene Symbol: 1700123O12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045185.1 | 5 | 555 | |||
| GenBank mRNA | AK007260.1 | - | 5 | 556 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY707359.1 | - | 5 | 554 | Chr. 4 - 10508031 - 10797801 [-] on Build GRCm38 | |

1700123O12Rik
Genomic Map